Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

804827

Sigma-Aldrich

MADN

Synonym(s):

2-Methyl-9,10-bis(naphthalen-2-yl)anthracene, 2-Methyl-9,10-di(2′-naphthyl)anthracene, 2-Methyl-9,10-di(2-naphthyl)anthracene, 2-Methyl-9,10-di(7-naphthalenyl)anthracene, 2-methyl-9,10-di-2-naphthalenylanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H24
CAS Number:
Molecular Weight:
444.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

>99% (HPLC)

form

solid

mp

253-258 °C

SMILES string

CC1=CC2=C(C3=CC4=CC=CC=C4C=C3)C5=CC=CC=C5C(C6=CC7=CC=CC=C7C=C6)=C2C=C1

InChI

1S/C35H24/c1-23-14-19-32-33(20-23)35(29-18-16-25-9-3-5-11-27(25)22-29)31-13-7-6-12-30(31)34(32)28-17-15-24-8-2-4-10-26(24)21-28/h2-22H,1H3

InChI key

HNWFFTUWRIGBNM-UHFFFAOYSA-N

Application

Fluorescent host material in high efficiency blue OLED devices.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and electroluminescence properties of highly efficient blue fluorescence emitters using dual core chromophores.
Beomjin K, et al.
Journal of Material Chemistry C, 1(3), 432-440 (2013)
Highly Efficient, Deep-Blue Doped Organic Light-Emitting Devices.
Lee MT, et al.
Advanced Materials, 17(20), 2493-2497 (2005)
Hybrid white organic light-emitting diodes with improved color stability and negligible efficiency roll-off based on blue fluorescence and yellow phosphorescence.
Xuehui W, et al.
Journal of Luminescence, 137, 59-63 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service