Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

804711

Sigma-Aldrich

Potassium 4-fluorobenzoyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4BF4KO
CAS Number:
Molecular Weight:
230.01
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

SMILES string

O=C(B(F)(F)F)C1=CC=C(F)C=C1.[K]

InChI

1S/C7H4BF4O.K.H/c9-6-3-1-5(2-4-6)7(13)8(10,11)12;;/h1-4H;;

InChI key

BDIWURUUSCKZDU-UHFFFAOYSA-N

General description

Potassium 4-fluorobenzoyltrifluoroborate belongs to the potassium acyltrifluoroborate (KAT) group. It can be synthesized from 1-(methoxy(4-fluorophenyl)methyl)-1H-benzotriazole.

Application

Potassium 4-fluorobenzoyltrifluoroborate may be used in the synthesis of MIDA (N-methyliminodiacetic acid) 4-fluorobenzoylboronate. It may also be used in the synthesis of potassium O-(4-benzoyltrifluoroborate)-O′-methylpolyethylene glycols of varying molecular weights.
Potassium Acyltrifluroborates (KAT′s) are bench, air, and moisture stable reagents for rapid, chemoselective amide bond formations with hydroxylamines. These amide bond forming reactions proceed under aqueous conditions, without the need for coupling reagents or protecting groups. This product was introduced in collaboration with the Bode Research Group.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and chemoselective ligations of MIDA acylboronates with O-Me hydroxylamines.
Noda H and Bode JW.
Chemical Science, 5(11), 4328-4332 (2014)
Hidetoshi Noda et al.
Journal of the American Chemical Society, 136(15), 5611-5614 (2014-04-02)
The identification of fast, chemoselective bond-forming reactions is one of the major contemporary challenges in chemistry. We show that chemoselective amide-forming ligations of potassium acyltrifluoroborates (KATs) and O-carbamoylhydroxylamines proceed in the presence of all unprotected functional groups with a second-order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service