About This Item
Recommended Products
form
powder
mp
178-183 °C (d)
functional group
phenyl
SMILES string
[K+].F[B-](F)(F)C(=O)Cc1ccccc1
InChI
1S/C8H7BF3O.K/c10-9(11,12)8(13)6-7-4-2-1-3-5-7;/h1-5H,6H2;/q-1;+1
InChI key
SEOSMCJOPUBHBF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Amide-Forming Ligation of Acyltrifluoroborates and Hydroxylamines in Water
Synthesis of Chemically and Configurationally Stable Monofluoro Acylboronates: Effect of Ligand Structure on their Formation, Properties, and Reactivities
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service