Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

804649

Sigma-Aldrich

(Ethylthio-trifluoroborate)-methane dimethyliminium

95%

Synonym(s):

Bode KAT Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11BF3NS
CAS Number:
Molecular Weight:
185.02
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

storage temp.

2-8°C

SMILES string

F[B-](F)(F)C(SCC)=[N+](C)C

InChI

1S/C5H11BF3NS/c1-4-11-5(10(2)3)6(7,8)9/h4H2,1-3H3

InChI key

FOEJEMWJAWZDLW-UHFFFAOYSA-N

Application

Reagent is a benchtop stable solid used in the preparation of potassium acyltrifluoroborates(KAT′s). Reported reaction conditions require 1.0 equiv. of n-BuLi(in hexane) at -78°C from the corresponding aryl/heteroaryl halide. Resulting KAT′s can be isolated without needing column chromatography.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service