Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

804576

Sigma-Aldrich

(1E)-4-Hydroxy-1-mesityl-4-methylpent-1-en-3-one

Synonym(s):

Bode Hydroxamic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O2
CAS Number:
Molecular Weight:
232.32
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

crystals

mp

75-77 °C

functional group

hydroxyl
ketone

SMILES string

O=C(C(C)(C)O)/C=C/C1=C(C)C=C(C)C=C1C

InChI

1S/C15H20O2/c1-10-8-11(2)13(12(3)9-10)6-7-14(16)15(4,5)17/h6-9,17H,1-5H3/b7-6+

InChI key

MZEPKOZFBZWACD-VOTSOKGWSA-N

Application

The following hydroxamic acid was shown by Bode and coworkers to assist in the kinetic resolution of cyclic amines through acylation of one enantiomer, which can be controlled using either the (R,S)- or (S,R)-Bode Kinetic Resolution Catalyst, both of which are also available through Sigma-Aldrich. 

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service