804096
N-(4-((1-methylcyclohexyl)amino) phenyl)acetamide
About This Item
form
powder
storage temp.
2-8°C
SMILES string
CC1(CCCCC1)NC2=CC=C(NC(C)=O)C=C2
InChI
1S/C15H22N2O/c1-12(18)16-13-6-8-14(9-7-13)17-15(2)10-4-3-5-11-15/h6-9,17H,3-5,10-11H2,1-2H3,(H,16,18)
InChI key
WUYRDZPQDNTNNI-UHFFFAOYSA-N
Application
related product
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service