803812
Mn3O4/reduced graphene oxide nanocomposite
10 mg/mL, dispersion in acetone
Synonym(s):
Mn3O4 decorated reduced graphene oxide, Mn3O4/RGO nanocomposite
About This Item
Recommended Products
description
Loading Density: 30%-70% (Mn3O4 nanocrystal) by TEM
Resistance: < 103 Ω/sq (reduced graphene oxide)
form
dispersion
composition
acetone, ~80%
Mn3O4 nanocrystal, 3-8%
reduced graphene oxide, 10-17%
concentration
10 mg/mL in acetone
thickness
0.5-1 nm , reduced graphene oxide
particle size
5-25 nm (Mn3O4 nanocrystal)
storage temp.
2-8°C
Application
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
59.0 °F
flash_point_c
15 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.
Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.
Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service