803790
Pd/reduced graphene oxide nanocomposite
10 mg/mL, dispersion in acetone
Synonym(s):
Pd decorated graphene oxide, Pd/RGO nanocomposite
About This Item
Recommended Products
description
Loading Density: 10%-50% (Pd nanocrystal) by TEM
Resistance: <103 Ω/sq (graphene)
form
dispersion
composition
acetone, ~80%
Pd nanocrystal, <5%
reduced graphene oxide, 5-20%
concentration
10 mg/mL
thickness
1-10 nm , graphene
particle size
5-50 nm (Pd nanocrystal)
storage temp.
2-8°C
Application
- Perfect for transmission electron microscopy imaging standard.
- A catalytic material to be used in fuel cells, CH4 gas sensor.
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
41.0 °F
flash_point_c
5 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.
Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service