803782
Pt/reduced graphene oxide nanocomposite
10 mg/mL, dispersion in acetone
Synonym(s):
Pt decorated reduced graphene oxide, Pt/RGO nanocomposite
About This Item
Recommended Products
description
Loading Density: 5%-30% (Pt nanocrystal) by TEM
Resistance: <103 Ω/sq (graphene)
form
dispersion
composition
acetone, ~80 wt. %
Pt nanocrystal, <5%
reduced graphene oxide, 5-20%
concentration
10 mg/mL in acetone
thickness
0.5-1 nm , reduced graphene oxide
particle size
2-5 nm (Pt nanocrystal)
storage temp.
2-8°C
Application
- Supercapacitor electrode materials.
- Filling materials for heating conduction.
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
59.0 °F
flash_point_c
15 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.
Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service