Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

803731

Sigma-Aldrich

Silica nanospheres

180 nm avg. part. size (TEM), 10 % (w/v) in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
SiO2
Molecular Weight:
60.08
UNSPSC Code:
12352300

form

nanospheres

concentration

10 % (w/v) in ethanol

avg. part. size

180 nm (TEM)

Looking for similar products? Visit Product Comparison Guide

Application

Traditionally the silica (SiO2) nanoparticles find applications as additives to drugs, cosmetics, printer toners, varnishes, and food etc. In recent years, their use has been extended to biomedical and biotechnological fields as biosensors, biomarkers, DNA and drug delivery agents etc.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

61.9 °F

flash_point_c

16.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service