Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

803324

Sigma-Aldrich

Sulfo-SIAB (sulfosuccinimidyl (4-iodoacetyl)aminobenzoate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O8N2SINa
Molecular Weight:
504.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

powder

mol wt

504.2

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C1CC(S(=O)([O-])=O)C(N1OC(C2=CC=C(NC(CI)=O)C=C2)=O)=O.[Na+]

InChI

1S/C13H11IN2O8S.Na/c14-6-10(17)15-8-3-1-7(2-4-8)13(20)24-16-11(18)5-9(12(16)19)25(21,22)23;/h1-4,9H,5-6H2,(H,15,17)(H,21,22,23);/q;+1/p-1

InChI key

HHSGWIABCIVPJT-UHFFFAOYSA-M

General description

SIAB is an amine- and sulfhydryl-reactive heterobifunctional crosslinker and Sulfo-SIAB is its water-soluble analog. These crosslinkers contain an amine-reactive N-hydroxysuccimide (NHS) ester and a sulfhyryl-reactive iodoacetyl group, and are often used for preparing enzyme conjugates or immunotoxins. Water-soluble and water-insoluble forms of NHS esters have essentially identical reactivity. Sulfo-SIAB is supplied as a sodium salt and is water-soluble to ~10mM. SIAB must be first dissolved in an organic solvent, such as DMSO or DMF, and then added to the aqueous reaction mixture. SIAB is lipophilic, membrane-permeable and does not possess a charged group.

Features and Benefits

  • Reactive groups: sulfo-NHS ester and iodoacetyl
  • Reactive toward: amino and sulfhydryl groups
  • Non-cleavable
  • Water-soluble (compare to SIAB)

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service