Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

803251

Sigma-Aldrich

Sulfo-KMUS (N-(κ-maleimidoundecanoyloxy) sulfosuccinimide ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25N2O9SNa
Molecular Weight:
480.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

powder

mol wt

480.47

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C1CC(S(=O)([O-])=O)C(N1OC(CCCCCCCCCCN2C(C=CC2=O)=O)=O)=O.[Na+]

InChI

1S/C19H26N2O9S.Na/c22-15-10-11-16(23)20(15)12-8-6-4-2-1-3-5-7-9-18(25)30-21-17(24)13-14(19(21)26)31(27,28)29;/h10-11,14H,1-9,12-13H2,(H,27,28,29);/q;+1/p-1

InChI key

MKNJJMHQBYVHRS-UHFFFAOYSA-M

General description

Sulfo-KMUS is a heterobifunctional crosslinker with N-hydroxysuccinimide (NHS) ester and maleimide groups that allow covalent conjugation of amine- and sulfhydryl-containing molecules. NHS esters react with primary amines at pH 7-9 to form amide bonds, while maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. In aqueous solutions, hydrolytic degradation of the NHS ester is a competing reaction whose rate increases with pH. The maleimide group is more stable than the NHS-ester group but will slowly hydrolyze and also lose its reaction specificity for sulfhydryls at pH values greater than 7.5. For these reasons, conjugation experiments involving this type of heterobifunctional crosslinker are usually performed at pH 7.2-7.5, with the NHS-ester (amine-targeted) reaction being accomplished before or simultaneous with the maleimide (sulfhydryl-targeted) reaction.

Features and Benefits

  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
  • Long aliphatic spacer arm compared to Sulfo-GMBS
  • Sulfo-NHS ester reacts with primary amines at pH 7-9, forming a stable amide bond
  • Maleimide reacts with -SH groups at pH 6.5-7.5, forming a stable thioether bond
  • Non-cleavable
  • Water-soluble

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenhui Chen et al.
The Journal of biological chemistry, 278(48), 48348-48356 (2003-09-16)
Heterobifunctional thiol to amine cross-linking agents were used to gain new insights on the dynamics and conformational factors governing the interaction between the cardiac Ca2+ pump (SERCA2a) and phospholamban (PLB). PLB is a small protein inhibitor of SERCA2a that reduces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service