Aquivion® PFSA ionomer membranes are melt-extruded films based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) F2C=CF-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. Following a film hydrolysis the perfluoropolymer′s functional groups are operative in their sulfonic acid form, SO3H.
Application
Aquivion® PFSA membranes are used for electrochemical applications such as, but not limited to, polymer electrolyte fuel cells, electro-deionization systems, ozone generators, water electrolyzers, hydrogen separators and compressors, redox flow batteries as well as pervaporation or gas humidification systems.
Packaging
Membranes are packed and sealed in a barrier protection pouch inside a cardboard box to prevent damage or deterioration. It is recommendable storing the product in a clean, humidity-controlled environment protected from direct sunlight or other sources of irradiation or heat. Sheet dimensions are based on product conditioned at 23°C and about 50% relative humidity. They may slightly vary upon exposure to different ambient conditions. Maximum temperature rating is 230°C, overheating will result in polymer degradation, discoloration and release of toxic volatile decomposition product.
Legal Information
Aquivion is a registered trademark of Syensqo Group
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.