Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

802638

Sigma-Aldrich

Aquivion® PW98

coarse powder, PFSA eq. wt. 980 g/mole SO3H

Synonym(s):

Aquivion® SO3H, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF2CF(OCF2CF2SO3H)]m[CF2CF2]n
CAS Number:
UNSPSC Code:
26111700
NACRES:
NA.23

form

coarse powder

Looking for similar products? Visit Product Comparison Guide

General description

Aquivion® PFSA PW series grades are coarse powders based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) CF2=CF2-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. The resulting perfluoropolymer′s functional groups are in their sulfonic acid form, SO3H.

Application

  • Aquivion®PW98 acid powder exhibits ion cation-exchange capacity and can be used as such under conditions where conventional exchange resins have shown insufficient chemical resistance.
  • Given the resin′s superacid functionality it can also be used either directly or as an ingredient of composite structures in the field of heterogeneous catalysis. This encompasses a wide range of organic synthesis and respective reaction mechanisms, as already described in public literature.

Legal Information

Aquivion is a registered trademark of Syensqo Group

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service