Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

802530

Sigma-Aldrich

Aquivion® P87S-SO2F

cylindrical pellets 2mm, PFSF eq. wt. (870 g/mole SO2F), contains CF3 polymer chain ends as stabilizer

Synonym(s):

Aquivion® SO2F-form, Perfluoro(3-oxa-4-pentenesulfonyl fluoride)-tetrafluoroethylene copolymer, Ethanesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF2CF(OCF2CF2SO2F)]m[CF2CF2]n
CAS Number:
UNSPSC Code:
26111700

Quality Level

form

pellets

eq. wt.

870g/mol±30g/mol SO2F

contains

CF3 polymer chain ends as stabilizer

Looking for similar products? Visit Product Comparison Guide

General description

Aquivion® P87S-SO2F are pellets based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) CF2=CF2-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. The resulting perfluoropolymer′s functional groups are in their sulfonyl fluoride form, SO2F. The polymer of this grade is chemically stabilized and therefore contains a maximized ratio of inert R-CF3 end-groups.

application

  • Aquivion® P87S pellets can be melt-pressed or extruded into plates, sheets or thin films down to about 50μ. The extruder shall be equipped with a standard three-zone metering screw devoid of barrier or mixing elements. Optimum temperature settings will depend on the configuration of the equipment but generally cover 100 to 300°C range. Optionally functional groups in the molded part are then hydrolyzed in baths or wet benches with a heated base (NaOH, KOH etc), reaction which may take minutes or hours, depending on thickness and conditions.
  • Aquivion® P87S pellets can be directly hydrolyzed as described - either completely or only on their outer surface. This will enable functional activity of the sulfonic acid / sulfonyl salt groups for ion-exchange operations or as acid catalyst.

Legal Information

Aquivion is a registered trademark of Syensqo Group

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service