Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

802506

Sigma-Aldrich

9,9′-(2,2′-Dimethyl[1,1′-biphenyl]-4,4′-diyl)bis-9H-carbazole

Synonym(s):

4,4′-Bis(9-carbazolyl)-2,2′-dimethylbiphenyl, CDBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H28N2
CAS Number:
Molecular Weight:
512.64
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

96% (HPLC)

form

powder

mp

96 °C

SMILES string

CC1=CC(N2C3=CC=CC=C3C4=C2C=CC=C4)=CC=C1C5=C(C)C=C(N6C7=C(C=CC=C7)C8=CC=CC=C68)C=C5

InChI

1S/C38H28N2/c1-25-23-27(39-35-15-7-3-11-31(35)32-12-4-8-16-36(32)39)19-21-29(25)30-22-20-28(24-26(30)2)40-37-17-9-5-13-33(37)34-14-6-10-18-38(34)40/h3-24H,1-2H3

InChI key

LTUJKAYZIMMJEP-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of neutral d10 coinage metal complexes with an anionic bidentate ligand in delayed fuorescence-type organic light-emitting diodes.
Osawa et al.
Journal of Material Chemistry C, 1(28), 4375-4383 (2013)
A series of CBP-derivatives as host materials for blue phosphorescent organic light-emitting diodes.
Schrogel P, et al.
Journal of Material Chemistry C, 21, 2266-2273 (2011)
Highly efficient blue phosphorescent and electroluminescent Ir(III) compounds.
Kang et al.
Journal of Material Chemistry C, 1(3), 441-450 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service