Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

799513

Sigma-Aldrich

Chiral cyclohexane diamine cycloimine cage

Synonym(s):

CC3-R, Cage 3-R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H84N12
CAS Number:
Molecular Weight:
1117.52
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

powder

storage temp.

2-8°C

SMILES string

C12/N=C/C3=CC(/C=N/C4CCCCC4/N=C/C5=CC(/C=N/C(CCCC6)C6/N=C/7)=CC(/C=N/C8CCCCC8/N=C/C9=CC(/C=N/C%10C(CCCC%10)/N=C/C%11=CC7=CC%12=C%11)=CC(/C=N/C1CCCC2)=C9)=C5)=CC(/C=N/C%13CCCCC%13/N=C\%12)=C3

InChI

1S/C72H84N12/c1-2-14-62-61(13-1)73-37-49-25-51-29-52(26-49)40-76-64-16-4-6-18-66(64)80-44-56-32-58-36-60(34-56)48-84-72-24-12-11-23-71(72)83-47-59-33-55(43-79-65-17-5-3-15-63(65)75-39-51)31-57(35-59)45-81-69-21-9-7-19-67(69)77-41-53-27-50(38-74-62)28-54(30-53)42-78-68-20-8-10-22-70(68)82-46-58/h25-48,61-72H,1-24H2/b73-37-,74-38+,75-39+,76-40+,77-41+,78-42+,79-43+,80-44+,81-45+,82-46+,83-47+,84-48+

InChI key

VIMCYPYFNMOHLL-YFWIPYMHSA-N

Application

CC3-R is shape persistent porous organic cage that can be used to separate gases and small molecules based on their size and shape.[1][2][3]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linjiang Chen et al.
Nature materials, 13(10), 954-960 (2014-07-21)
The separation of molecules with similar size and shape is an important technological challenge. For example, rare gases can pose either an economic opportunity or an environmental hazard and there is a need to separate these spherical molecules selectively at
Tamoghna Mitra et al.
Nature chemistry, 5(4), 276-281 (2013-03-21)
The energy-efficient separation of chemical feedstocks is a major sustainability challenge. Porous extended frameworks such as zeolites or metal-organic frameworks are one potential solution to this problem. Here, we show that organic molecules, rather than frameworks, can separate other organic
Tomokazu Tozawa et al.
Nature materials, 8(12), 973-978 (2009-10-27)
Porous materials are important in a wide range of applications including molecular separations and catalysis. We demonstrate that covalently bonded organic cages can assemble into crystalline microporous materials. The porosity is prefabricated and intrinsic to the molecular cage structure, as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service