Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

799114

Sigma-Aldrich

2-(Methacryloyloxy)ethyl [3-(triethoxysilyl)propyl]carbamate

contains ≤1000 ppm phenothiazine as inhibitor, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H31NO7Si
CAS Number:
Molecular Weight:
377.51
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

95%

form

liquid

contains

≤1000 ppm phenothiazine as inhibitor

storage temp.

2-8°C

SMILES string

CC(C(OCCOC(NCCC[Si](OCC)(OCC)OCC)=O)=O)=C

InChI

1S/C16H31NO7Si/c1-6-22-25(23-7-2,24-8-3)13-9-10-17-16(19)21-12-11-20-15(18)14(4)5/h4,6-13H2,1-3,5H3,(H,17,19)

InChI key

HBFXZOMNDOKZCW-UHFFFAOYSA-N

Application

Hydrophilic siloxysilylalkylacrylamide monomers are used in the manufacture of contact lens and medical devices.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service