799092
Graphene nanoplatelets
1 mg/mL, dispersion in H2O, avg. no. of layers, 5 ‑ 7
Synonym(s):
GNP dispersion, Graphene dispersion
About This Item
Recommended Products
description
Surfactant type: Anionic Surfactant
form
dispersion in H2O
feature
avg. no. of layers 5 ‑ 7
composition
Graphene, 0.1 wt. %
Graphene+Surfactant, 0.2 wt. %
Water, 99.8 wt. %
Water, 99.9 wt. %
concentration
1 mg/mL
sheet resistance
10 (+/-5) Ω/sq (for a 25 μm film)
Looking for similar products? Visit Product Comparison Guide
General description
Surfactant Type: Anionic Surfactant
Sheet Resistantce: 10 (+/-5) ohm/sq for a 25 μm film
Raman D/G and D/D′: 0.28 and 5.0 (The 2D band shape indicates the presence of few-layer graphene flakes with an average of 5-7 atomic layers)
Application
- Graphene (nano)composite materials
- Conductive coatings
- Conductive Inks
- Energy Storage
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.
Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.
Graphene's unique properties spark interdisciplinary interest; its honeycomb structure offers electrical, optical, and mechanical marvels.
Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service