Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

799033

Sigma-Aldrich

Poly(ethylene glycol) methyl ether-block-poly(ε-caprolactone)

PEG average Mn 5,000, PCL average Mn 25,000, PDI ≤1.6

Synonym(s):

PEG-PCL, PEG-b-PCL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C2H4O)nO(C6H10O2)mH
UNSPSC Code:
12162002
NACRES:
NA.23

description

Tm: 63-68°C

form

solid

mol wt

PCL average Mn 25,000
PEG average Mn 5,000
apparent mol wt 20000-30000 (PCL)
apparent mol wt 4000-6000 (PEG)

PDI

≤1.6

storage temp.

2-8°C

application

Biodegradable polymer for drug delivery.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service