798940
Trovitch Mn-Precatalyst
Synonym(s):
((Ph2PPr)PDI)Mn
About This Item
Recommended Products
form
powder
reaction suitability
core: manganese
reagent type: catalyst
mp
>200 °C
storage temp.
2-8°C
SMILES string
C/C(C1=CC=CC(/C(C)=N/CCCP(C2=CC=CC=C2)C3=CC=CC=C3)=N1)=N\CCCP(C4=CC=CC=C4)C5=CC=CC=C5.[Mn]
InChI
1S/C39H41N3P2.Mn/c1-32(40-28-16-30-43(34-18-7-3-8-19-34)35-20-9-4-10-21-35)38-26-15-27-39(42-38)33(2)41-29-17-31-44(36-22-11-5-12-23-36)37-24-13-6-14-25-37;/h3-15,18-27H,16-17,28-31H2,1-2H3;/b40-32+,41-33+;
InChI key
HGQXTZFURCAPHA-PVFFQWOCSA-N
General description
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The Trovitch laboratory is working to develop first-row transition metal catalysts that can be used to achieve a variety of organic transformations. The first complexes from this group made available through Sigma-Aldrich are the hydrosilylation catalysts, (Ph2PPrPDI)Mn and (Ph2PPrDI)Ni. Although these compounds are formally zerovalent, both feature a reduced chelate that coordinates to the metal center beyond its redox non-innocent core.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service