Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798827

Sigma-Aldrich

(S)-N-Butyl-1-[(2-hydroxybenzyl)-L-valyl]pyrrolidine-2-carboxamide

Synonym(s):

Yoon Syn-[2+2]-Photocycloaddition Ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H33N3O3
CAS Number:
Molecular Weight:
375.51
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

liquid

storage temp.

2-8°C

SMILES string

OC1=C(CN[C@@H](C(C)C)C(N2[C@H](C(NCCCC)=O)CCC2)=O)C=CC=C1

InChI

1S/C21H33N3O3/c1-4-5-12-22-20(26)17-10-8-13-24(17)21(27)19(15(2)3)23-14-16-9-6-7-11-18(16)25/h6-7,9,11,15,17,19,23,25H,4-5,8,10,12-14H2,1-3H3,(H,22,26)/t17-,19-/m0/s1

InChI key

YETNAJXRRQTUPV-HKUYNNGSSA-N

General description

(S)-N-Butyl-1-[(2-hydroxybenzyl)-L-valyl]pyrrolidine-2-carboxamide is a secondary amine ligand. It act as a Lewis acid co-catalyst for [2+2] cycloaddition on complexation with Eu(OTf)3 (europium(III)triflate).[1]

Application

Chiral ligand for the asymmetric [2+2] cycloaddition of enones by photoredox catalysis.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juana Du et al.
Science (New York, N.Y.), 344(6182), 392-396 (2014-04-26)
In contrast to the wealth of catalytic systems that are available to control the stereochemistry of thermally promoted cycloadditions, few similarly effective methods exist for the stereocontrol of photochemical cycloadditions. A major unsolved challenge in the design of enantioselective catalytic

Related Content

Yoon lab exploits visible light photochemistry for unique chemical transformations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service