Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798517

Sigma-Aldrich

Titanium dioxide

22 nm & >150 nm particle size (BET), > 95% anatase (XRD), colloidal paste for opaque film

Synonym(s):

Nanocrystalline TiO2, TiO2 paste, Titania paste

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

description

Metals purity: ≤ 11 ppm Fe, Na and K (ICP)
Rheology at shear rate: 0.3189 at 1000cm-1 (Pa.s)
Rheology at shear rate: 646.2 at 0.01 cm-1 (Pa.s)

form

paste (nanocrystalline colloid)

concentration

16 wt. %

surface area

50-60 m2/g , BET

particle size

22 nm & >150 nm (BET)

pore size

34-36 nm average pore size (BET)
55-60 % porosity (BET)

pH

<1

conductivity

77.6 mS

Looking for similar products? Visit Product Comparison Guide

Application

Titanium dioxide (TiO2) is used in the creation of photoelectrodes for the production of dye sensitized, perovskite and other solid state solar cells. Our TiO2 pastes allow deposition of titania films with large surface to volume ratio. Select transparent TiO2 paste for applications that require a transparent sintered titania film and use opaque TiO2 paste for applications that do not require transparency. The opaque TiO2 paste contains larger scattering TiO2 particles.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service