Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798312

Sigma-Aldrich

3-(2-Ethylhexyl)thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20S
CAS Number:
Molecular Weight:
196.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

refractive index

n/D 1.494

bp

150-152 °C/7.5 Torr

density

0.915 g/mL at 25 °C

SMILES string

CCC(CCCC)CC1=CSC=C1

InChI

1S/C12H20S/c1-3-5-6-11(4-2)9-12-7-8-13-10-12/h7-8,10-11H,3-6,9H2,1-2H3

InChI key

HWMQCIZYXLAJKM-UHFFFAOYSA-N

Application

3-(2-Ethylhexyl)thiophene is a 3-alkylthiophene monomer, which can be used in the formation of conjugated highly regio-regular block polymers for applications in organic field effect transistors (OFETs), and organic photovoltaics (OPVs).[1][2][3][4]
Organic semiconducting polymer or small molecule precursor.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Influence of intermolecular interactions of electron donating small molecules on their molecular packing and performance in organic electronic devices
Kim, Ki-Hyun; Yu, Hojeong; Kang, Hyunbum; et al.
Journal of Material Chemistry A, 1(46), 14538-14547 (2013)
Nanostructure formation in poly (3-hexylthiophene-block-3-(2-ethylhexyl) thiophene) s
Zhang Y, et al.
Macromolecules, 42(18), 7008-7015 (2009)
3, 6-Dialkylthieno [3, 2-b] thiophene moiety as a soluble and electron donating unit preserving the coplanarity of photovoltaic low band gap copolymers
Biniek L, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 50(9), 1861-1868 (2012)
Impact of the Alkyl Side Chains on the Optoelectronic Properties of a Series of Photovoltaic Low-?Band-?Gap Copolymers
Biniek, Laure; Fall; Sadiara, Chochos; Christos L., Anokhin; et al.
Macromolecules, 43(23), 9779-9786 (2010)
Studies on 3-(2-ethylhexyl)thiophene polymers
Somanathan, Narayanasastri; Radhakrishnan, Sarangapani; Thelakkat, Mukundan; Schmidt, Hans W.
Macromolecular Materials and Engineering, 287(4), 236-242 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service