Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798290

Sigma-Aldrich

2,5-Dibromo-3-(2-ethylhexyl)thiophene

97% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Br2S
Molecular Weight:
354.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97% (GC)

form

liquid

SMILES string

BrC1=C(CC(CC)CCCC)C=C(Br)S1

InChI

1S/C12H18Br2S/c1-3-5-6-9(4-2)7-10-8-11(13)15-12(10)14/h8-9H,3-7H2,1-2H3

InChI key

DFUZDCATNROZQR-UHFFFAOYSA-N

Application

Organic semiconducting polymer or small molecule precursor.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-Hwan Lee et al.
Journal of the American Chemical Society, 135(47), 17695-17698 (2013-11-15)
Nanostar and nanonetwork crystals were prepared from fully conjugated poly(3-(2-ethylhexyl)thiophene)-block-polythiophene (P3EHT-b-PT) via a simple INCP process. The structural conformation of the nanocrystals was investigated in detail, revealing that with an increase in the block length of PT, the morphology of
Tuning Polythiophene Crystallization through Systematic Side Chain Functionalization
Ho, Victor; Boudouris, Bryan W.; Segalman, Rachel A.
Macromolecules, 43(19), 7895-7899 (2010)
Studies on 3-(2-ethylhexyl)thiophene polymers
Somanathan, Narayanasastri; Radhakrishnan, Sarangapani; Thelakkat, Mukundan; Schmidt, Hans W.
Macromolecular Materials and Engineering, 287(4), 236-242 (2002)
Shrayesh N Patel et al.
Nano letters, 12(9), 4901-4906 (2012-07-31)
We report on the synthesis and morphology of a block copolymer, poly(3-(2'-ethylhexyl)thiophene)-b-poly(ethylene oxide) (P3EHT-b-PEO), that conducts both electrons and ions. We show that in the melt state the P3EHT-b-PEO chains self-assemble to produce traditional nanoscale morphologies such as lamellae and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service