Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798274

Sigma-Aldrich

Poly(dimethylsiloxane), monomethacrylate terminated

contains ≤500 ppm phenothiazine as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H15O3[C2H6OSi]nC2H6Si-R
UNSPSC Code:
12162002

form

liquid

contains

≤500 ppm phenothiazine as inhibitor
50-500 ppm Phenothiazine as inhibitor

refractive index

n20/D 1.408

transition temp

flash point >230 °F

density

0.971 g/mL

storage temp.

2-8°C

Application

UV curable PDMS based monomer for silicone hydrogel application in medical devices.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service