Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797839

Sigma-Aldrich

Poly(D,L-lactide-block-acrylic acid)

PDLA Mn 18,000, PAA Mn 11,000, PDI ≤1.6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H25S3(C3H4O2)xC6H9NO(C3H4O2)yH
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

Quality Level

mol wt

PAA Mn 11,000
PDLA Mn 18,000

PDI

≤1.6

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

This diblock copolymer can be used to self-assemble into a drug delivery vehicle. This particular block ratio and molecular weight forms cylindrical micelles.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service