Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797804

Sigma-Aldrich

Poly(D,L-lactide), 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentonate terminated

average Mn 10,000, PDI <1.4

Synonym(s):

PDLLA RAFT terminated, RAFT-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H35NOS3(C3H4O2)n
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

average Mn 10,000

PDI

<1.4

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization

Legal Information

Sold for research purposes only: See sigma-aldrich.com/raftlicense. Patents: WO98/01478; WO99/311444.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan A Mendenhall et al.
Nature microbiology, 5(12), 1464-1471 (2020-09-09)
Anthrax lethal toxin (LT), produced by Bacillus anthracis, comprises a receptor-binding moiety, protective antigen and the lethal factor (LF) protease1,2. Although LF is known to cleave mitogen-activated protein kinase kinases (MEKs/MKKs) and some variants of the NLRP1 inflammasome sensor, targeting
Pierre Maudens et al.
Journal of controlled release : official journal of the Controlled Release Society, 276, 102-112 (2018-03-11)
This study aimed to formulate nanocrystal-polymer particles (NPPs) containing the potent p38α/β MAPK inhibitor PH-797804 (PH-NPPs) and to test their extended-release properties over months in comparison to those of conventional PH microparticles for the intra-articular treatment of inflammatory and mechanistic
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service