Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797790

Sigma-Aldrich

Poly(D,L-lactide), 4-cyano-4-[(dodecylsulfanylthiocarbonyl)sulfanyl]pentonate terminated

average Mn 20,000, PDI <1.4

Synonym(s):

PDLLA RAFT terminated, RAFT-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H35NOS3(C3H4O2)n
UNSPSC Code:
12352100
NACRES:
NA.23

form

crystals

mol wt

average Mn 20,000

PDI

<1.4

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization. Chain Transfer Agent (CTA).
Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Articles

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service