Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797502

Sigma-Aldrich

1-Benzyl-6-tosyl-1,6-diazaspiro[3.3]heptan-3-one

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O3S
CAS Number:
Molecular Weight:
356.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

storage temp.

2-8°C

SMILES string

O=C1CN(CC2=CC=CC=C2)C13CN(S(C4=CC=C(C)C=C4)(=O)=O)C3

InChI

1S/C19H20N2O3S/c1-15-7-9-17(10-8-15)25(23,24)21-13-19(14-21)18(22)12-20(19)11-16-5-3-2-4-6-16/h2-10H,11-14H2,1H3

InChI key

NIUURRMQZVBZSY-UHFFFAOYSA-N

Application

Spirocyclic building block is part of a series of advanced angular[3.3]heptanes developed by Carreira and coworkers.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service