797480
1-Benzyl-6-tosyl-1,6-diazaspiro[3.3]heptan-3-amine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H23N3O2S
CAS Number:
Molecular Weight:
357.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
(powder or crystal or solid)
Storage temp.
2-8°C
SMILES string
NC1CN(CC2=CC=CC=C2)C13CN(S(C4=CC=C(C)C=C4)(=O)=O)C3
InChI
1S/C19H23N3O2S/c1-15-7-9-17(10-8-15)25(23,24)22-13-19(14-22)18(20)12-21(19)11-16-5-3-2-4-6-16/h2-10,18H,11-14,20H2,1H3
Inchi Key
SMOIAYPLVUKPDW-UHFFFAOYSA-N
Application
Spirocyclic building block is part of a series of advanced angular[3.3]heptanes developed by Carreira and coworkers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service