797146
Iron oxide (II,III) magnetic nanoparticles solution
5 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL Fe in H2O, dispersion
Synonym(s):
Fe NP COOH, FexOy
About This Item
Recommended Products
form
dispersion
nanoparticles
concentration
5 mg/mL Fe in H2O
magnetization
>25 emu/g (at room temperature, under 4500 Oe)
avg. part. size
5 nm (TEM)
density
~1 g/mL at 25 °C
functional group
carboxylic acid
storage temp.
2-8°C
Application
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Prof. Yadong Yin discusses various synthesis methods of magnetite nanocrystals and their applications in different fields.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service