All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H10BNO2
CAS Number:
Molecular Weight:
187.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
assay
95%
form
powder
storage temp.
2-8°C
SMILES string
Cc1ccc2ncccc2c1B(O)O
InChI
1S/C10H10BNO2/c1-7-4-5-9-8(3-2-6-12-9)10(7)11(13)14/h2-6,13-14H,1H3
InChI key
CNTWHKAWJCAEIJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service