Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

796824

Sigma-Aldrich

Thiamine nitrate

95%

Synonym(s):

Vitamin B1 nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N5O4S
CAS Number:
Molecular Weight:
327.36
Beilstein/REAXYS Number:
3857790
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

95%

form

solid

color

faint beige to beige
white to yellow

mp

374-392 °C

SMILES string

[O-][N+]([O-])=O.CC1=NC=C(C[N+]2=CSC(CCO)=C2C)C(N)=N1

InChI

1S/C12H17N4OS.NO3/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;2-1(3)4/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);/q+1;-1

InChI key

UIERGBJEBXXIGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service