796514
Silver nanoplates
resonant wavelength, 980 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized
About This Item
Recommended Products
form
nanoplates
storage condition
protect from light
concentration
0.02 mg/mL (in water with 5 mM sodium borate buffer)
density
1.003 g/mL
resonant wavelength
980 nm
functional group
PVP
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 2
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service