796352
Silver nanospheres
100 nm avg. part. size, 0.02 mg/mL in water, PEG functionalized
About This Item
form
nanospheres
concentration
0.02 mg/mL in water
avg. part. size
100 nm
density
1.0011 g/mL at 25 °C
λmax
485 nm
functional group
PEG
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 2
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service