Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795828

Sigma-Aldrich

3,4-Difluorobenzylphosphonic acid

Synonym(s):

3,4-F2BPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7F2O3P
CAS Number:
Molecular Weight:
208.10
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

SMILES string

OP(CC1=CC=C(F)C(F)=C1)(O)=O

InChI

1S/C7H7F2O3P/c8-6-2-1-5(3-7(6)9)4-13(10,11)12/h1-3H,4H2,(H2,10,11,12)

InChI key

RBWZNPGUZZKJHV-UHFFFAOYSA-N

Application

Tuning of surface energy, wetting and work function of metal oxides.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organosulphur Phosphorus Acid Compounds. Part 7. Preparation and Analytical Identification of Difluorobenzylphosphono-Sulfonic Acids
Montoneri, E.;
Phosphorus, Sulfur, and Silicon and the Related Elements, 134, 99-100 (1998)
Peter J Hotchkiss et al.
Accounts of chemical research, 45(3), 337-346 (2011-10-21)
Transparent metal oxides, in particular, indium tin oxide (ITO), are critical transparent contact materials for applications in next-generation organic electronics, including organic light emitting diodes (OLEDs) and organic photovoltaics (OPVs). Understanding and controlling the surface properties of ITO allows for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service