Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795798

Sigma-Aldrich

p-Aminobenzylphosphonic acid

Synonym(s):

4-Aminobenzylphosphonic acid, ABPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10NO3P
CAS Number:
Molecular Weight:
187.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

98%

form

solid

SMILES string

Nc1ccc(CP(O)(O)=O)cc1

InChI

1S/C7H10NO3P/c8-7-3-1-6(2-4-7)5-12(9,10)11/h1-4H,5,8H2,(H2,9,10,11)

InChI key

NEKHKXMBGWNTOO-UHFFFAOYSA-N

Application

Tuning of surface energy, wetting and work function of metal oxides.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 18F-labelled stilbenes from 4-[18F]fluorobenz-aldehyde using the Horner?Wadsworth?Emmons reaction
Gester, S.;
Journal of Labelled Compounds & Radiopharmaceuticals, 50, 105-113 (2007)
Peter J Hotchkiss et al.
Accounts of chemical research, 45(3), 337-346 (2011-10-21)
Transparent metal oxides, in particular, indium tin oxide (ITO), are critical transparent contact materials for applications in next-generation organic electronics, including organic light emitting diodes (OLEDs) and organic photovoltaics (OPVs). Understanding and controlling the surface properties of ITO allows for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service