Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795593

Sigma-Aldrich

3-Fluorobenzylphosphonic acid

Synonym(s):

3-FBPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8FO3P
CAS Number:
Molecular Weight:
190.11
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

95%

form

powder

SMILES string

OP(CC1=CC(F)=CC=C1)(O)=O

InChI

1S/C7H8FO3P/c8-7-3-1-2-6(4-7)5-12(9,10)11/h1-4H,5H2,(H2,9,10,11)

InChI key

FEASAQQGBIZVJR-UHFFFAOYSA-N

Application

Tuning of surface energy, wetting, and work function of metal oxides.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myeng Chan Hong et al.
Bioorganic & medicinal chemistry, 18(22), 7724-7730 (2010-07-14)
Fluorescence probes that can detect Aβ (β-amyloid peptide) plaque are important tools for diagnosis of Alzheimer's disease (AD), and 4-N-methylamino-4'-hydroxystilbene (SB-13) is one of the promising candidate molecules. We report here the synthesis of SB-13 derivatives that consist of various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service