Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795372

Sigma-Aldrich

Naphtho[1,2-c:5,6-c′]bis[1,2,5]thiadiazole

>98% (HPLC)

Synonym(s):

NT, NTz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4N4S2
CAS Number:
Molecular Weight:
244.30
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

>98% (HPLC)

form

solid

SMILES string

C12=NSN=C1C(C=CC3=NSN=C34)=C4C=C2

InChI

1S/C10H4N4S2/c1-3-7-10(14-15-11-7)6-2-4-8-9(5(1)6)13-16-12-8/h1-4H

InChI key

VHHDKHOBNKXVNU-UHFFFAOYSA-N

Application

NTz contains two fused 1, 2, 5-thiadiazole rings that lower the band gap; enhance the interchain packing; and improve the charge mobility of the resulting polymer. The NT-based polymer PBDT-DTNT exhibited considerably better photovoltaic performance with a power conversion efficiency (PCE) of 6.00% .[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ming Wang et al.
Journal of the American Chemical Society, 133(25), 9638-9641 (2011-06-03)
Donor-acceptor conjugated polymers PBDT-DTBT and PBDT-DTNT, based on 2,1,3-benzothiadiazole (BT) and naphtho[1,2-c:5,6-c]bis[1,2,5]thiadiazole (NT), have been designed and synthesized for polymer solar cells. NT contains two fused 1,2,5-thiadiazole rings that lower the band gap, enhance the interchain packing, and improve the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service