Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

795186

Sigma-Aldrich

5-Bromopyridine-2-boronic acid MIDA ester

95% (HPLC)

Synonym(s):

MIDA021

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BBrN2O4
CAS Number:
Molecular Weight:
312.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95% (HPLC)

form

solid

mp

220-225 °C (decomposition)

functional group

bromo

SMILES string

BrC1=CC=C(B2OC(CN(C)CC(O2)=O)=O)N=C1

InChI

1S/C10H10BBrN2O4/c1-14-5-9(15)17-11(18-10(16)6-14)8-3-2-7(12)4-13-8/h2-4H,5-6H2,1H3

InChI key

RSJASKKCIPMBEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The 2-substituted pyridinyl boronic acid has classically been a challenging motif to install, due to instability of the boronic acid. The MIDA Boronate now enables a highly effecient means of installing the 2-pyridyl motif through Suzuki-Miyuara coupling.

A General Solution for the 2-Pyridyl Problem

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service