Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

794430

Sigma-Aldrich

3-O-Acryloyl-1,2:5,6-bis-O-isopropylidene-D-glucofuranose

99%

Synonym(s):

3-O-Acryloyl-1,2:5,6-di-O-(1-methylethylidene)-α-D-glucofuranose, protected glucofuranose acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O7
CAS Number:
Molecular Weight:
314.33
UNSPSC Code:
12162002
PubChem Substance ID:

assay

99%

form

powder

mp

76.9 °C

storage temp.

2-8°C

SMILES string

CC1(C)O[C@]2([H])[C@@H](OC(C=C)=O)[C@]([C@]3([H])OC(C)(C)OC3)([H])O[C@]2([H])O1

InChI

1S/C15H22O7/c1-6-9(16)18-11-10(8-7-17-14(2,3)20-8)19-13-12(11)21-15(4,5)22-13/h6,8,10-13H,1,7H2,2-5H3/t8-,10-,11+,12-,13-/m1/s1

InChI key

HMCNQVMKOONNJT-KABOQKQYSA-N

Application

This monomer can be used to make macromolecules containing carbohydrate moieties. Controlled Radical Polymerization, such as NMP,[1] RAFT, or ATRP,[2] is commonly employed to synthesize well-defined glycopolymers.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gotz, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 40(20), 3379-3391 (2002)
Muthukrishnan, et al.
Macromolecules, 38(1), 9-18 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service