Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

794414

Sigma-Aldrich

HRu(bmpi)(PPh3)2

Synonym(s):

Szymczak Active Dehydrogenation Precatalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H47N5P2Ru
CAS Number:
Molecular Weight:
953.02
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

170 °C (decomposition)

storage temp.

2-8°C

SMILES string

[H][Ru](N1/C(C2=C(C=CC=C2)/C1=N\C3=CC=CC(C)=N3)=N/C4=CC=CC(C)=N4)(P(C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7)P(C8=CC=CC=C8)(C9=CC=CC=C9)C%10=CC=CC=C%10

InChI

1S/C20H16N5.2C18H15P.Ru.H/c1-13-7-5-11-17(21-13)23-19-15-9-3-4-10-16(15)20(25-19)24-18-12-6-8-14(2)22-18;2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3-12H,1-2H3;2*1-15H;;/q-1;;;-1;/p+2

InChI key

MNBCXKMRCBBDOA-UHFFFAOYSA-P

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service