Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

794406

Sigma-Aldrich

Ru(bmpi)(PPh3)Cl

95%

Synonym(s):

Szymczak Air-Stable Dehydrogenation Precursor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H31ClN5PRu
CAS Number:
Molecular Weight:
725.18
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

powder

mp

345 °C (decomposition)

storage temp.

2-8°C

SMILES string

Cl[Ru](N1/C(C2=C(C=CC=C2)/C1=N\C3=CC=CC(C)=N3)=N/C4=CC=CC(C)=N4)P(C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7

InChI

1S/C20H16N5.C18H15P.ClH.Ru/c1-13-7-5-11-17(21-13)23-19-15-9-3-4-10-16(15)20(25-19)24-18-12-6-8-14(2)22-18;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3-12H,1-2H3;1-15H;1H;/q-1;;;+1

InChI key

HLSBTSDBHMUZSH-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service