Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

794201

Sigma-Aldrich

Neopentyl(t-Bu)2P Pd G3

95%

Synonym(s):

[(Di-tert-butylneopentylphosphine)-2-(2-aminobiphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H42NO3PPdS
CAS Number:
Molecular Weight:
586.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

powder

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

mp

178-183 °C

functional group

phosphine

SMILES string

NC(C=CC=C1)=C1C2=C([Pd]OS(C)(=O)=O)C=CC=C2.CC(C)(C)P(C(C)(C)C)CC(C)(C)C

InChI

1S/C13H29P.C12H10N.CH4O3S.Pd/c1-11(2,3)10-14(12(4,5)6)13(7,8)9;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h10H2,1-9H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

GWXFWQLHGGYESC-UHFFFAOYSA-M

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service