Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

794163

Sigma-Aldrich

IBiox[(−)-menthyl] HOTf

95%

Synonym(s):

(1S,2S,2′′S,5R,5′′R,7′S)-2,2′′-Diisopropyl-5,5′′-dimethyl-2′H,8′H-dispiro[cyclohexane-1,3′-imidazo[4,3-b:5,1-b′]bis(oxazole)-7′,1′′-cyclohexan]-4′-ium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H41F3N2O5S
CAS Number:
Molecular Weight:
550.67
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

optical activity

[α]22/D +31.0°, c = 0.5% in chloroform

mp

216-221 °C

SMILES string

CC(C)[C@@H]1C[C@H](C)CC[C@]21COC3=C4[N+]([C@]5(C[C@H](C)CC[C@H]5C(C)C)CO4)=CN32.O=S([O-])(C(F)(F)F)=O

InChI

1S/C25H41N2O2.CHF3O3S/c1-16(2)20-8-7-19(6)12-25(20)14-29-23-22-26(15-27(23)25)24(13-28-22)10-9-18(5)11-21(24)17(3)4;2-1(3,4)8(5,6)7/h15-21H,7-14H2,1-6H3;(H,5,6,7)/q+1;/p-1/t18-,19-,20+,21+,24-,25-;/m1./s1

InChI key

MXNKPJQKMTUZSH-JSWGGJIYSA-M

Application

Chiral NHC ligand was reported to ligate to palladium(II) precatalysts for highly stereoselective transformations.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service