Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

793760

Sigma-Aldrich

cis-8,11,14-Eicosatrienoic acid-1-13C

99 atom % 13C, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH=CHCH2CH=CH(CH2)613COOH
Molecular Weight:
307.48
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

assay

97% (CP)

form

liquid

mass shift

M+1

storage temp.

−20°C

InChI

1S/C21H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23/h7-8,10-11,13-14H,2-6,9,12,15-20H2,1H3,(H,22,23)/b8-7-,11-10-,14-13-/i21+1

InChI key

MVSHFHIWJPEGNV-CCQVIMDTSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service