Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

793701

Sigma-Aldrich

5-Trifluoromethyl-2-pyridylboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10BF3N2O4
CAS Number:
Molecular Weight:
302.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

207-212 °C

SMILES string

FC(F)(F)C1=CN=C(C=C1)B2OC(CN(C)CC(O2)=O)=O

InChI

1S/C11H10BF3N2O4/c1-17-5-9(18)20-12(21-10(19)6-17)8-3-2-7(4-16-8)11(13,14)15/h2-4H,5-6H2,1H3

InChI key

QFNWEZKEFLNTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Pyridyl substituted MIDA boronate is one of several offerings that overcome several problems other 2-pyridyl substituted boronates have, e.g. protodeborylation, long-term storage, insufficient reactivity with aryl chlorides.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service