793698
4-Chloro-2-methylphenoxyacetic acid-(phenyl-13C6)
99 atom % 13C, 98% (CP)
About This Item
isotopic purity
99 atom % 13C
assay
98% (CP)
form
solid
application(s)
agriculture
mass shift
M+6
SMILES string
C[13C]1=[13C](OCC(O)=O)[13CH]=[13CH][13C](Cl)=[13CH]1
InChI
1S/C9H9ClO3/c1-6-4-7(10)2-3-8(6)13-5-9(11)12/h2-4H,5H2,1H3,(H,11,12)/i2+1,3+1,4+1,6+1,7+1,8+1
InChI key
WHKUVVPPKQRRBV-BOCFXHSMSA-N
Related Categories
Packaging
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service