Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

793426

Sigma-Aldrich

4-Fluorophenyl-α-MIDA-boryl aldehyde

95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13BFNO5
CAS Number:
Molecular Weight:
293.06
UNSPSC Code:
12352103
PubChem Substance ID:

assay

95% (HPLC)

mp

168-173 °C

storage temp.

2-8°C

SMILES string

O=C1OB2(C(C3=CC=C(F)C=C3)C([H])=O)OC(C[N](C)2C1)=O

InChI

1S/C13H13BFNO5/c1-16-6-12(18)20-14(16,21-13(19)7-16)11(8-17)9-2-4-10(15)5-3-9/h2-5,8,11H,6-7H2,1H3

InChI key

JJWSMIFASSANSV-UHFFFAOYSA-N

General description

4-Fluorophenyl-α-MIDA-boryl aldehyde is an α-MIDA (α-N-methyliminodiacetyl) boryl aldehyde derivative.

Application

This boronic acid derivative is in a series of the first known stable α-boryl aldehyde species. Through the employment of the MIDA-protected boronic ester, this building block has been shown as a precursor to the synthesis of unnatural amino acids.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey D St Denis et al.
Organic letters, 17(23), 5764-5767 (2015-11-21)
Herein, we describe the rhodium-catalyzed C-H amination reaction of 1,2-boryl sulfamate esters derived from amphoteric α-boryl aldehydes. Depending on the substitution pattern of the boryl sulfamate ester, a diverse range of five- or six-membered ring heterocycles are accessible using this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service